Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Thioridazine hydrochloride
GREENER_CHOICESDP

Catalog No. 307050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-705-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-705-0 Supplier Tocris Bioscience™ Supplier No. 3070/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dopamine receptor antagonist; antipsychotic

Dopamine receptor antagonist that displays antipsychotic activity. Exhibits anticancer activity in cervical, endometrial and breast cancer cells; triggers apoptosis by targeting the PI 3-K/Akt/mTOR/p70 S6K pathway. Induces G1 cell cycle arrest. Selective inducer of cancer stem cell (CSC) differentiation.

Chemical Identifiers

CAS 130-61-0
Molecular Formula C21H27ClN2S2

Specifications

Quantity 50 mg
Formula Weight 407.04
Purity >99%
Molecular Weight (g/mol) 407.031
Chemical Name or Material Thioridazine hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.