Learn More
THOC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 153161AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0143
Only null left
Add to Cart
Description
Description
THOC6 belongs to the WD repeat THOC6 family. It contains 7 WD repeats. The function of the THOC6 protein remains unknown.
Specifications
Specifications
THOC6
Polyclonal
Unconjugated
THOC6
fSAP35, THO complex subunit 6 homolog, THOC6, WDR58
Rabbit
Antigen Affinity Chromatography
RUO
386612, 79227, 79228
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5U4D9, Q6AY87, Q86W42
Thoc6
THOC6 Fusion Protein Ag7561
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.