Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THOC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | THOC6 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0143
![]() ![]() |
Proteintech Group Inc
153161AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
THOC6 belongs to the WD repeat THOC6 family. It contains 7 WD repeats. The function of the THOC6 protein remains unknown.Specifications
THOC6 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5U4D9, Q6AY87, Q86W42 | |
386612, 79227, 79228 | |
THOC6 Fusion Protein Ag7561 | |
Primary | |
-20°C | |
THOC6 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
fSAP35, THO complex subunit 6 homolog, THOC6, WDR58 | |
Thoc6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title