Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomsen-Friedenreich Antigen Antibody (A68-B/A11) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB121644 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121644 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121644 Supplier Novus Biologicals Supplier No. NBP311557
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thomsen-Friedenreich Antigen Monoclonal antibody specifically detects Thomsen-Friedenreich Antigen in Human, Mouse, Rat samples. It is validated for ELISA

Specifications

Antigen Thomsen-Friedenreich Antigen
Applications ELISA
Classification Monoclonal
Clone A68-B/A11
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution ELISA 1-5 ug/ml
Formulation 10 mM PBS
Host Species Mouse
Immunogen Neuraminidase-treated human red blood cells
Purification Method Bioreactor concentrate
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.