Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ThPOK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142375
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142375 Supplier Invitrogen™ Supplier No. PA5142375
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

ZFP67 is an early growth response gene that encodes a zinc finger-containing transcription factor that binds to the promoter regions of type I collagen genes (e.g., COL1A1; MIM 120150) and has a role in development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ThPOK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Zbtb7b
Gene Accession No. O15156
Gene Alias CKROX; c-KROX; DKFZp686G01254; hcKROX; Krueppel-related zinc finger protein cKrox; RP11-307C12.9; T helper-inducing POZ/Krueppel factor; T-helper-inducing POZ/Krueppel-like factor; Thpok; Vzinc finger and BTB domain containing 7B; ZBTB15; ZBTB7B; ZFP67; Zfp-67; zinc finger and BTB domain containing 15; zinc finger and BTB domain containing 7B; Zinc finger and BTB domain-containing protein 15; Zinc finger and BTB domain-containing protein 7B; zinc finger protein 67; Zinc finger protein 67 homolog; zinc finger protein 857B; Zinc finger protein Th-POK; ZNF857B
Gene Symbols Zbtb7b
Host Species Goat
Immunogen Peptide with sequence C-DSQDKLVRKRRSQ.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51043
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.