Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRAP3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224477SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224477SS 0.025 mg
NBP224477 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224477SS Supplier Novus Biologicals Supplier No. NBP224477SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

THRAP3 Polyclonal specifically detects THRAP3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen THRAP3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation PBS + 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9Y2W1
Gene Alias FLJ22082, MGC133082, MGC133083, thyroid hormone receptor associated protein 3, Thyroid hormone receptor-associated protein complex 150 kDa component, thyroid hormone receptor-associated protein, 150 kDa subunit, Trap150, TRAP150thyroid hormone receptor-associated protein 3
Gene Symbols THRAP3
Host Species Rabbit
Immunogen A portion of amino acids 930-955 of human Thyroid hormone receptor associated protein 3 was used as the immunogen
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9967
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), horse, cow, dog, mouse, monkeyand chimpanzee, 93% homologous in chicken, 78% homologous in xenopus and 73% homologous in rat and opossum.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.