Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THRAP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | THRAP3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.1-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224477SS
![]() |
Novus Biologicals
NBP224477SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224477
![]() |
Novus Biologicals
NBP224477 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
THRAP3 Polyclonal specifically detects THRAP3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
THRAP3 | |
Western Blot 0.1-1 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
Polyclonal | |
Rabbit | |
Signal Transduction, Transcription Factors and Regulators | |
PBS + 0.05% BSA with 0.05% Sodium Azide | |
FLJ22082, MGC133082, MGC133083, thyroid hormone receptor associated protein 3, Thyroid hormone receptor-associated protein complex 150 kDa component, thyroid hormone receptor-associated protein, 150 kDa subunit, Trap150, TRAP150thyroid hormone receptor-associated protein 3 | |
THRAP3 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a), horse, cow, dog, mouse, monkeyand chimpanzee, 93% homologous in chicken, 78% homologous in xenopus and 73% homologous in rat and opossum. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Primate | |
Q9Y2W1 | |
9967 | |
A portion of amino acids 930-955 of human Thyroid hormone receptor associated protein 3 was used as the immunogen | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title