Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombin-antithrombin (TAT) complexes Antibody (22), PerCP, Novus Biologicals™
SDP

Catalog No. NB343280 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343280 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343280 Supplier Novus Biologicals Supplier No. NBP241381PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thrombin-antithrombin (TAT) complexes Monoclonal antibody specifically detects Thrombin-antithrombin (TAT) complexes in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen Thrombin-antithrombin (TAT) complexes
Applications Immunohistochemistry
Classification Monoclonal
Clone 22
Conjugate PerCP
Dilution Immunohistochemistry
Formulation PBS
Host Species Mouse
Immunogen Thrombin-antithrombin (TAT) complexes
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.