Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Antibody (4D8.2C6), PE, Novus Biologicals™
SDP

Catalog No. NB006989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB006989 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB006989 Supplier Novus Biologicals Supplier No. NBP236496PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Thrombomodulin/BDCA-3
Applications Flow Cytometry
Classification Monoclonal
Clone 4D8.2C6
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Gene Symbols THBD
Host Species Rat
Immunogen Two synthetic peptides made to internal portions of the mouse Thrombomodulin protein (between residues 25-150 and 400-550) [UniProt P07204]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.