Learn More
Thrombomodulin/BDCA-3 Antibody (6C8.2F9), DyLight 405, Novus Biologicals™
Shop All R&D Systems Products

Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Thrombomodulin/BDCA-3 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 6C8.2F9 |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | P07204 |
| Gene Alias | BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM |
| Gene Symbols | THBD |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.