Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Antibody (THBD/1591) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254490200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254490200 200 μg
NB254490100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254490200 Supplier Novus Biologicals Supplier No. NBP254490200UG

Mouse Monoclonal Antibody

Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Thrombomodulin/BDCA-3
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone THBD/1591
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Gene Symbols THBD
Host Species Mouse
Immunogen Recombinant human Thrombomodulin protein fragment (aa69-194) (exact sequence is proprietary)
Molecular Weight of Antigen 100 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7056
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.