Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Mouse anti-Human, Clone: QBEND/40, Novus Biologicals™
SDP

Catalog No. NB10064982 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064982 0.1 mL
NB017676 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064982 Supplier Novus Biologicals Supplier No. NB10064982

Mouse Monoclonal Antibody

Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy.

Specifications

Antigen Thrombomodulin/BDCA-3
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone QBEND/40
Conjugate Unconjugated
Dilution Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:100, Immunohistochemistry-Frozen 1:10-1:100, Electron Microscopy
Gene Accession No. P07204
Gene Alias BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Gene Symbols THBD
Host Species Mouse
Immunogen Human endothelium (thrombomodulin)
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7056
Test Specificity Recognizes all human endothelium tested. Preliminary evidence suggests that this recognizes Thrombomodulin, but does not inhibit Thrombomodulin dependent activation of protein C.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.