Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombopoietin/THPO Antibody (444), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB333216 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333216 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333216 Supplier Novus Biologicals Supplier No. NBP290449MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Thrombopoietin/THPO Monoclonal antibody specifically detects Thrombopoietin/THPO in Mouse samples. It is validated for ELISA

Specifications

Antigen Thrombopoietin/THPO
Applications ELISA
Classification Monoclonal
Clone 444
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Thrombopoietin/THPO (Accession#: NP_033405.1, Met 1-Thr 356)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7066
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.