Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombopoietin/THPO Antibody (MM0566-1Z3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP211922IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211922IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211922IR Supplier Novus Biologicals Supplier No. NBP211922IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Western Blot.

Specifications

Antigen Thrombopoietin/Tpo
Applications Western Blot
Classification Monoclonal
Clone MM0566-1Z3
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF
Gene Symbols THPO
Host Species Mouse
Immunogen Human recombinant Thrombopoietin/THPO
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7066
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.