Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-1 Rabbit anti-Human, Mouse, Clone: S06-5F3, Novus Biologicals™
SDP

Catalog No. NB165409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165409 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165409 Supplier Novus Biologicals Supplier No. NBP315101100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Thrombospondin-1 Monoclonal antibody specifically detects Thrombospondin-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Thrombospondin-1
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S06-5F3
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias THBS, THBS-1, thrombospondin 1, thrombospondin-1p180, TSP1thrombospondin-1, TSPTSP-1
Host Species Rabbit
Immunogen Recombinant protein of human Thrombospondin-1 (Uniprot#: P07996)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7057
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.