Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thromboxane synthase Antibody (OTI2C1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB274505APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274505APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274505APC Supplier Novus Biologicals Supplier No. NBP274505APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thromboxane synthase Monoclonal specifically detects Thromboxane synthase in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Thromboxane synthase
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2C1
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CYP5A1family 5, subfamily A, polypeptide 1, CYP5FLJ52771, EC 5.3.99.5, GHOSAL, platelet, cytochrome P450, subfamily V, thromboxane A synthase 1 (platelet), thromboxane A synthase 1 (platelet, cytochrome P450, family 5, subfamily A), thromboxane A synthase 1 (platelet, cytochrome P450, subfamily V), thromboxane-A synthase, TXA synthase
Gene Symbols TBXAS1
Host Species Mouse
Immunogen Full length recombinant protein of human Thromboxane synthase (NP_001052) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 6916.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.