Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-1 Antibody (A65M), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP105185G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105185G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105185G Supplier Novus Biologicals Supplier No. NBP105185G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thrombospondin-1 Monoclonal antibody specifically detects Thrombospondin-1 in Human, Bovine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen Thrombospondin-1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone A65M
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias THBS, THBS-1, thrombospondin 1, thrombospondin-1p180, TSP1thrombospondin-1, TSPTSP-1
Gene Symbols THBS1
Host Species Mouse
Immunogen Extracellular matrix material from cultured bovine corneal endothelial cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7057
Test Specificity Highly specific for the low-calcium (0.1 mmol/L) conformation of human TSP1. There is no evidence of cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin).
Target Species Human, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.