Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THTPA Antibody (OTI13E3), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274511A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274511A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274511A700 Supplier Novus Biologicals Supplier No. NBP274511AF700

Mouse Monoclonal Antibody

THTPA Monoclonal specifically detects THTPA in Human samples. It is validated for Western Blot.

Specifications

Antigen THTPA
Applications Western Blot
Classification Monoclonal
Clone OTI13E3
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.6.1.28, MGC2652, thiamine triphosphatase, thiamine-triphosphatase, THTP, THTPASE
Gene Symbols THTPA
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-230 of human THTPA (NP_077304) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79178.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.