Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THTPA Antibody (OTI13E3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP274511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274511 Supplier Novus Biologicals Supplier No. NBP274511

Mouse Monoclonal Antibody

THTPA Monoclonal specifically detects THTPA in Human samples. It is validated for Western Blot.

Specifications

Antigen THTPA
Applications Western Blot
Classification Monoclonal
Clone OTI13E3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias EC 3.6.1.28, MGC2652, thiamine triphosphatase, thiamine-triphosphatase, THTP, THTPASE
Gene Symbols THTPA
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-230 of human THTPA (NP_077304) produced in E.coli.
Molecular Weight of Antigen 25.4 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79178
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.