Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THUMPD1 Antibody (OTI1H6), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274512A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274512A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274512A48 Supplier Novus Biologicals Supplier No. NBP274512AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

THUMPD1 Monoclonal specifically detects THUMPD1 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen THUMPD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1H6
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686C1054, FLJ20274, THUMP domain containing 1, THUMP domain-containing protein 1
Gene Symbols THUMPD1
Host Species Mouse
Immunogen Full length recombinant protein of human THUMPD1 (NP_060206) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55623.0
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.