Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidine glycol Rabbit anti-Human, Clone: B29, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00 - $687.50
Specifications
Antigen | Thymidine glycol |
---|---|
Clone | B29 |
Concentration | 1 mg/ml |
Dilution | ELISA, Immunohistochemistry 1:100 |
Applications | ELISA, Immunohistochemistry |
Description
Thymidine glycol Monoclonal specifically detects Thymidine glycol in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Thymidine glycol | |
1 mg/ml | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Thymidine glycol polymer | |
Primary | |
Store at -20°C. |
B29 | |
ELISA, Immunohistochemistry 1:100 | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 50% Glycerol | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title