Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidine Kinase 1 Antibody (6F6), Novus Biologicals™
SDP

Catalog No. NB298405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298405 100 μL
NB298404 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298405 Supplier Novus Biologicals Supplier No. NBP326286100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Thymidine Kinase 1 Monoclonal antibody specifically detects Thymidine Kinase 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry

Specifications

Antigen Thymidine Kinase 1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6F6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Thymidine Kinase 1 [UniProt P04183]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7083
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.