Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidine Kinase 1 Antibody (6F6), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Thymidine Kinase 1 |
---|---|
Clone | 6F6 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Thymidine Kinase 1 Monoclonal antibody specifically detects Thymidine Kinase 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
Thymidine Kinase 1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK2 | |
A synthesized peptide derived from Human Thymidine Kinase 1 [UniProt P04183] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6F6 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7083 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title