Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidylate Synthase Antibody (TYMS/1884), Biotin, Novus Biologicals™
SDP

Catalog No. NB389784 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389784 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389784 Supplier Novus Biologicals Supplier No. NBP308882B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thymidylate Synthase Monoclonal antibody specifically detects Thymidylate Synthase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Thymidylate Synthase
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TYMS/1884
Conjugate Biotin
Formulation PBS
Gene Alias EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422
Host Species Mouse
Immunogen Recombinant human thymidylate synthase protein fragment (around aa 60-174) (exact sequence is proprietary) (Uniprot: P04818)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 7298
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.