Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Thymidylate Synthase Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | Thymidylate Synthase |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119461
|
Invitrogen™
PA5119461 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121547
|
Invitrogen™
PA5121547 |
20 μL |
Each for $209.50
|
|
|||||
Description
Thymidylate Synthase Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Thymidylate Synthase (TS) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.Specifications
Thymidylate Synthase | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
TYMS | |
HST422; MGC88736; OK/SW-cl.29; Thymidylate synthase; thymidylate synthetase; TMS; TS; Tsase; Tyms; tysy | |
TYMS | |
Primary | |
Antigen affinity chromatography |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P04818 | |
7298 | |
Recombinant protein Thymidylate Synthetase. The antigen corresponds to amino acid range 36-313 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title