Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymine Dimer Antibody (H3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB6001141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001141 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001141 Supplier Novus Biologicals Supplier No. NB6001141
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thymine Dimer Monoclonal antibody specifically detects Thymine Dimer in Human, Nonspecies specific, Chicken samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot, In Situ Hybridization (ISH).

Specifications

Antigen Thymine Dimer
Applications ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot, Southern Blot
Classification Monoclonal
Clone H3
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Dot Blot 0.5-1 ug/ml, Southern Blot 0.5-1 ug/ml
Host Species Mouse
Immunogen tetra nucleotide containing cyclobutane thymine dimer (GTTG) conjugated to chicken gamma globulin.
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Test Specificity Monoclonal Anti-Thymine Dimer reacts specifically with the (5'-6') cyclobutane type of homothymine or thyminecytosine heterodimers. The antibody reacts with thymine dimers in single-stranded DNA, and has a lower affinity for the dimer in short oligonucleotides (a tail of minimum 10-20 thymine residues is required for efficient labeling of oligonucleotide probes). The product enables a sensitive and non-radioactive method for labeling, detection, and quantification of DNA fragments using ELISA, competitive ELISA, immunocytochemistry (laser-scan microscopy) and Southern immunoblotting.
Target Species Human, Non-species specific, Chicken
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.