Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals cis-4,7,10,13,16,19-Docosahexaenoic acid, 98%

Catalog No. AC427641000
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass Bottle

Catalog No. AC427641000

Supplier: Thermo Scientific Chemicals 427641000

Only null left
Add to Cart

CAS: 6217-54-5 | C22H32O2 | 328.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.49
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O

Specifications

Melting Point -44°C
Color Colorless to Yellow
Density 0.9500g/mL
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2CHCH)6CH2CH2COOH
Quantity 100 mg
Specific Gravity 0.95
Merck Index 15, 3443
Synonym cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
Formula Weight 328.49
Percent Purity 98%
Physical Form Oil
Chemical Name or Material cis-4,7,10,13,16,19-Docosahexaenoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.