Learn More
Thermo Scientific Chemicals Thymol Blue, ACS
Description
pH indicator: 1.2 (pink) to 2.8 (yellow) to 9.6 (blue). Thymol Blue is useful as a pH indicator. It is a component of universal indicator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 223°C (decomposition) |
| Merck Index | 14,9400 |
| Solubility Information | Soluble in alcohol and dilute alkali solutions; Insoluble in water . |
| Formula Weight | 466.6 |
| Grade | ACS |
| Quantity | 2 g |
| Physical Form | Crystalline |
| Chemical Name or Material | Thymol Blue |
Safety and Handling
| EINECSNumber | 200-973-3 |
| RTECSNumber | XP2575000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.