Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymol Blue Indicator, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Melting Point | 221°C to 224°C |
| UN Number | NONH for all modes of transport |
| Beilstein | 368036 |
| Formula Weight | 466.59g/mol |
| Packaging | Glass Bottle |
| Visual Transition Interval | 1.2 to 2.8 (red to yellow) |
| Density | 0.979g/cm3 |
| Quantity | 25 g |
| Physical Form | Powder |
| Chemical Name or Material | Thymol Blue |
Safety and Handling
| EINECSNumber | 200-973-3 |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction