Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymopoietin/LAP2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB164350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164350 Supplier Novus Biologicals Supplier No. NBP281736B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Thymopoietin/LAP2 Polyclonal antibody specifically detects Thymopoietin/LAP2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Thymopoietin/LAP2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CMD1T, lamina-associated polypeptide 2, LAP2PRO0868, LEM domain containing 4, LEMD4, MGC61508, thymopoietin, Thymopoietin isoform alpha, Thymopoietin, isoforms beta/gamma, Thymopoietin-related peptide isoform alpha, Thymopoietin-related peptide isoforms beta/gamma, TP, TP alpha, TP beta/gamma, TPRP isoform alpha, TPRP isoforms beta/gamma
Host Species Rabbit
Immunogen Thymopoietin antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human Thymopoietin. The immunogen is located within amino acids 80 - 130 of Thymopoietin. Amino Acid Squence: CVGRKATKKTDKPRQEDKDD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7112
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.