Learn More
THYN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 158671AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0541
Only null left
Add to Cart
Description
Description
THYN1, also named as THY28, is a 28-32 kDa protein which located mainly in the nucleus. It appears to correlate with induction of apoptosis.
Specifications
Specifications
THYN1
Polyclonal
Unconjugated
THYN1
HSPC144, MDS012, My0054, MY105, THY28, THY28KD, thymocyte nuclear protein 1, Thymocyte protein Thy28, THYN1
Rabbit
Antigen Affinity Chromatography
RUO
29087, 300470, 77862
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P3E0, Q91YJ3, Q9P016
THYN1
THYN1 Fusion Protein Ag8481
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.