Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THYN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | THYN1 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0541
![]() ![]() |
Proteintech Group Inc
158671AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
THYN1, also named as THY28, is a 28-32 kDa protein which located mainly in the nucleus. It appears to correlate with induction of apoptosis.Specifications
THYN1 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6P3E0, Q91YJ3, Q9P016 | |
29087, 300470, 77862 | |
THYN1 Fusion Protein Ag8481 | |
Primary | |
-20°C | |
THYN1 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HSPC144, MDS012, My0054, MY105, THY28, THY28KD, thymocyte nuclear protein 1, Thymocyte protein Thy28, THYN1 | |
THYN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title