Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroglobulin Antibody (5E6cc), Biotin
SDP

Catalog No. NB1108083B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1108083B 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1108083B Supplier Novus Biologicals Supplier No. NB1108083B

Mouse Monoclonal Antibody

Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Antigen Thyroglobulin
Applications ELISA
Classification Monoclonal
Clone 5000000
Conjugate Biotin
Dilution ELISA, Sandwich ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias AITD3TGN, TDH3, Tg, thyroglobulin
Gene Symbols TG
Host Species Mouse
Immunogen Thyroglobulin from human thyroid gland.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7038
Test Specificity This does not interfere with anti-thyroglobulin autoantibodies.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.