Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroglobulin Antibody (rTGB/4744), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB387665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387665 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387665 Supplier Novus Biologicals Supplier No. NBP308264AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thyroglobulin Monoclonal antibody specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen Thyroglobulin
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rTGB/4744
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias AITD3TGN, TDH3, Tg, thyroglobulin
Host Species Mouse
Immunogen Human thyroid follicular cells
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7038
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.