Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid Peroxidase Antibody (313), DyLight 680
SDP

Catalog No. NB070822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB070822 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB070822 Supplier Novus Biologicals Supplier No. NBP290193FR

Rabbit Monoclonal Antibody

Thyroid Peroxidase Monoclonal specifically detects Thyroid Peroxidase in Human samples. It is validated for ELISA.

Specifications

Antigen Thyroid Peroxidase
Applications ELISA
Classification Monoclonal
Clone 313
Conjugate DyLight 680
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Thyroid Peroxidase (P07202-1; Met1-Arg846,257Ser/Ala,725Pro/Thr).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7173
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.