Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid receptor-interacting protein 13 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB362649 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362649 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362649 Supplier Novus Biologicals Supplier No. NBP204046AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Thyroid receptor-interacting protein 13 Polyclonal antibody specifically detects Thyroid receptor-interacting protein 13 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Thyroid receptor-interacting protein 13
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias 16E1BP, 16E1-BP, HPV16 E1 protein binding protein, Human papillomavirus type 16 E1 protein-binding protein, thyroid hormone receptor interactor 13HPV16 E1 protein-binding protein, thyroid receptor interacting protein 13, thyroid receptor-interacting protein 13, TR-interacting protein 13, TRIP-13
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 382 and 432 of human Thyroid Hormone Receptor Interactor 13 using the numbering given in entry NP_004228.1 (GeneID 9319).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9319
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.