Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroxine Antibody (XM212), Novus Biologicals™
SDP

Catalog No. NB1107978 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1107978 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1107978 Supplier Novus Biologicals Supplier No. NB1107978
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thyroxine Monoclonal antibody specifically detects Thyroxine in Nonspecies specific samples. It is validated for ELISA.

Specifications

Antigen Thyroxine
Applications ELISA
Classification Monoclonal
Clone XM212
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS (pH 7.4) with 0.05% Sodium Azide
Gene Alias T4
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified thyroxine (Sigma T-2501) conjugated with BSA.
Purification Method Ion exchange chromatography
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This reacts with conjugate T4-BSA and with free thyroxine in ELISA. Cross-reactivity with triiodothyronine is less than 1%. The reactivity of this with L-Thyroxine is 100 % and with D-Thyroxine 30 %.
Target Species Non-species specific, Mouse, Rat, Mammalia
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.