Learn More
Description
Specifications
Specifications
| Antigen | TIA1 |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | TIA1/1313 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular) |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein |
| Gene Symbols | TIA1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
