Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIA1 Mouse anti-Human, Clone: TIA1/1313, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | TIA1 |
---|---|
Clone | TIA1/1313 |
Dilution | Western Blot 1-2ug/ml, Flow Cytometry 0.5-1ug/million cells in 0.1ml, SDS-Page, Immunofluorescence 1-2ug/ml |
Applications | Western Blot, Flow Cytometry, SDS-Page, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247642B
![]() |
Novus Biologicals
NBP2476420.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
TIA1 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Western Blotting.Specifications
TIA1 | |
Western Blot 1-2ug/ml, Flow Cytometry 0.5-1ug/million cells in 0.1ml, SDS-Page, Immunofluorescence 1-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
7072 | |
Recombinant human TIA1 fragment of 102 amino acid residues (aa279-380) | |
Primary | |
Store at 4C. | |
40 kDa |
TIA1/1313 | |
Western Blot, Flow Cytometry, SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein | |
TIA1 | |
IgG κ | |
Protein A purified | |
TIA-1 (T-cell intracytoplasmic antigen) is a cytoplasmic granule-associated protein, expressed in lymphocytes processing cytolytic potential. TIA-1 is a member of an RNA-binding protein family and possesses nucleolytic activity against cytotoxic lymphocyte (CTL) target cells. It has been suggested that this protein may be involved in the induction of apoptosis as it preferentially recognizes poly(A) homopolymers and induces DNA fragmentation in CTL targets. The major granule-associated species is a 15kDa protein thought to be derived from the carboxyl terminus of the 40kDa product by proteolytic processing. TIA1 antibody labels cytotoxic T cells and natural killer cells (NK cells). It is also expressed in T-cell lymphoma, large granular lymphocyte (LGL) leukemia and hairy cell leukemia. TIA1 expression in T-cell malignancies may help in differentiating LGL leukemia (high expression) from T-cell lymphocytosis and other T-cell diseases (low expression). TIA1 may also be used to label tum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title