Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIAF1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB183603 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183603 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183603 Supplier Novus Biologicals Supplier No. NBP282022AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIAF1 Polyclonal antibody specifically detects TIAF1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TIAF1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias 12 kDa TGF-beta-1-induced antiapoptotic factor, KIAA0216, MAJN, molecule associated with Jak-3 N-terminal, MYO18A, MYSPDZ, SPR210, TGFB1-induced apoptotic factor 1, TGF-beta-1-induced antiapoptotic factor 1
Host Species Rabbit
Immunogen TIAF1 antibody was raised against a 16 amino acid peptide near the amino terminus of human TIAF1 . The immunogen is located within the first 50 amino acids of TIAF1. Amino Acid Squence: CAGEESRVQVLKNEVRRG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9220
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.