Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIAL1 Antibody (6E3), Biotin, Novus Biologicals™
SDP

Catalog No. NB151647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151647 Supplier Novus Biologicals Supplier No. NBP311834B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIAL1 Monoclonal antibody specifically detects TIAL1 in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow

Specifications

Antigen TIAL1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone 6000
Conjugate Biotin
Formulation PBS
Gene Alias aging-associated gene 7 protein, MGC33401, TCBP, T-cluster binding protein, TIA1 cytotoxic granule-associated RNA binding protein-like 1, TIA1 cytotoxic granule-associated RNA-binding protein-like 1, TIA-1 related protein, TIA-1-related nucleolysin, TIA-1-related protein, TIARnucleolysin TIAR
Host Species Mouse
Immunogen GST-tagged recombinant human TIAL1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 7073
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.