Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIAM1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292435 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292435 Supplier Novus Biologicals Supplier No. NB1002301PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIAM1 Polyclonal specifically detects TIAM1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen TIAM1
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias FLJ36302, human T-lymphoma invasion and metastasis inducing TIAM1 protein, 10TIAM-1, T-cell lymphoma invasion and metastasis 1, T-lymphoma invasion and metastasis-inducing protein 1
Host Species Rabbit
Immunogen Maps to a region between residues 1550 and the C-terminus (residue 1591) of human T-cell lymphoma invasion and metastasis 1 using the numbering given in entry NP_003244.1 (GeneID 7074)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7074
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.