Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ticlopidine Hydrochloride

Catalog No. p-7082384
Click to view available options
Quantity:
5 g
25 g

Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines | CAS: 53885-35-1 | C14H14ClNS·HCl

Ticlopidine hydrochloride is a selective P2Y12 receptor antagonist. It inhibits ADP-induced platelet aggregation and displays antithrombotic activity following oral administration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53885-35-1
Molecular Formula C14H14ClNS·HCl
Synonym 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride

Specifications

Quantity 25 g
Description White
For Use With (Application) Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
Formula Weight 300.25g/mol
Product Type Signal Transduction Reagent
Form Powder
EINECSNumber 258-837-4
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.