Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TICRR Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343494 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343494 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343494 Supplier Novus Biologicals Supplier No. NBP241283MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TICRR Polyclonal antibody specifically detects TICRR in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen TICRR
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias C15orf42, Chromosome 15 Open Reading Frame 42, SLD3, SLD3 Homolog, SLD3 Homolog (S. Cerevisiae), TOPBP1-Interacting Checkpoint And Replication Regulator, TOPBP1-Interacting Replication-Stimulating Protein, TopBP1-Interacting, Replication-Stimulating Protein, Treslin
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human TICRR. The immunogen is located within amino acids 1840 - 1890 of TICRR. Amino Acid Squence: FQGKTPSSQSKDPRDEDVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 90381
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.