Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie-2 Antibody (83715), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB363206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363206 Supplier Novus Biologicals Supplier No. FAB3131M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen Tie-2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 83715
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Tie-2, Ala23-Lys745, Accession # AAA61139
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7010
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.