Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie-2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB338189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338189 Supplier Novus Biologicals Supplier No. NBP169753APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tie-2 Polyclonal antibody specifically detects Tie-2 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Tie-2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human TIE2 protein (within residues 250-400). [Swiss-Prot Q02763]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7010
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.