Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie-2 Antibody (tek9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP118614A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP118614A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP118614A30 Supplier Novus Biologicals Supplier No. NBP118614AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA.

Specifications

Antigen Tie-2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone tek9
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Gene Symbols TEK
Host Species Mouse
Immunogen Recombinant human soluble extracellular TIE-2
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7010.0
Test Specificity The monoclonal antibody will detect native human TIE-2/tek in ELISA experiments and on the surface of different human cell types.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.