Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tie-2 Antibody (tek9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018375 0.02 mg
NBP118614 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018375 Supplier Novus Biologicals Supplier No. NBP1186140.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA, CyTOF-ready.

Specifications

Antigen Tie-2
Applications Flow Cytometry, Western Blot, Immunofluorescence, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone tek9
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 - 5 ug/ml, Flow Cytometry 2 - 5 ug/ml, ELISA 1 - 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 5 ug/ml, Sandwich ELISA, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1
Gene Symbols TEK
Host Species Mouse
Immunogen recombinant human Jagged-1
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7010
Test Specificity The monoclonal antibody will detect native human TIE-2/tek in ELISA experiments and on the surface of different human cell types.
Reconstitution Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.