Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie-2 Antibody (tek9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$214.28 - $591.65
Specifications
| Antigen | Tie-2 |
|---|---|
| Clone | tek9 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 - 5 ug/ml, Flow Cytometry 2 - 5 ug/ml, ELISA 1 - 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 5 ug/ml, Sandwich ELISA, CyTOF-ready |
| Applications | Flow Cytometry, Western Blot, Immunofluorescence, ELISA, Immunohistochemistry, Immunocytochemistry |
Description
Tie-2 Monoclonal specifically detects Tie-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Sandwich ELISA, CyTOF-ready.Specifications
| Tie-2 | |
| LYOPH | |
| Flow Cytometry, Western Blot, Immunofluorescence, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Protein Kinase | |
| angiopoietin-1 receptor, CD202B, CD202b antigen, EC 2.7.10, EC 2.7.10.1, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, TEK tyrosine kinase, endothelial, TIE-2, TIE2CD202b, Tunica interna endothelial cell kinase, Tyrosine-protein kinase receptor TEK, Tyrosine-protein kinase receptor TIE-2, venous malformations, multiple cutaneous and mucosal, VMCM, VMCM1 | |
| TEK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| tek9 | |
| Western Blot 2 - 5 ug/ml, Flow Cytometry 2 - 5 ug/ml, ELISA 1 - 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 5 ug/ml, Sandwich ELISA, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 7010 | |
| IgG1 | |
| Protein G purified | |
| The monoclonal antibody will detect native human TIE-2/tek in ELISA experiments and on the surface of different human cell types. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title