Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIF1 alpha Antibody (OTI2D9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP274514R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274514R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274514R Supplier Novus Biologicals Supplier No. NBP274514R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TIF1 alpha Monoclonal specifically detects TIF1 alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TIF1 alpha
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2D9
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24
Gene Symbols TRIM24
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 706-1016 of human TIF1 alpha (NP_003843) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 8805.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.